2-Amino-5-chloropyrimidine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Chloropyrimidin-2-ylamine, 5-Chloro-2-pyrimidinamine, 5-Chlor-2-pyrimidinamin, 5-chlorpyrimidin-2-amin

001651-5G 001651-25G 001651-100G
101782-188EA 15.38 USD
101782-188 101782-190 101782-192
2-Amino-5-chloropyrimidine ≥98%
2-Amino-5-chloropyrimidine

Hazard code(s): Irritant

Formula: C₄H₄ClN₃
MW: 129.55 g/mol
Melting Pt: 235 °C
MDL Number: MFCD00014604
CAS Number: 5428-89-7
EINECS: 226-581-2

Order Now

Specification Test Results

Melting point 235°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR