4-Bromo-3-chlorophenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

001648-1G 001648-5G
101782-178EA 63.08 USD
101782-178 101782-180
4-Bromo-3-chlorophenyl isothiocyanate ≥98%
4-Bromo-3-chlorophenyl isothiocyanate

Hazard code(s): Irritant, Lachrymator

Formula: C₇H₃BrClNS
MDL Number: MFCD00041071
CAS Number: 32118-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR