4-Bromo-2-chloro-1-iodobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-3-chloro-4-iodobenzene

001645-25G 001645-100G
101782-170EA 36.92 USD
101782-170 101782-172
4-Bromo-2-chloro-1-iodobenzene ≥98%
4-Bromo-2-chloro-1-iodobenzene

Hazard code(s): Irritant

Formula: C₆H₃BrClI
MW: 317.35 g/mol
Melting Pt: 33…35 °C
MDL Number: MFCD00079709
CAS Number: 31928-47-9

Order Now

Specification Test Results

Melting point 33-35°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR