4-Chloro-2-(trifluoromethyl)aniline ≥97% for synthesis
Supplier: Matrix Scientific
Synonyms:
2-Amino-5-chloro-α,α,α-trifluorotoluene, 2-Amino-5-chloro-α,α,α-trifluortoluene, 2-Amino-5-chlorobenzotrifluoride, 4-Chloro-α,α,α-trifluoro-o-toluidine
001641-100G
001641-500G
101782-164EA
53.85
USD
101782-164
101782-166
4-Chloro-2-(trifluoromethyl)aniline ≥97% for synthesis
4-Chloro-2-(trifluoromethyl)aniline
Hazard code(s): Irritant
Formula:
C₇H₅ClF₃N Boiling Pt: 66…67 °C (3mm) Melting Pt: 8…9 °C Density: 1.38 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007841 CAS Number: 445-03-4 EINECS: 207-151-3 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 66-67°C/3mm |
Melting point | 8-9°C |
Density | 1.386 |
ND20 | 1.507 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...