3-Bromo-4-chlorobenzotrifluoride ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Bromo-2-chloro-5-(trifluoromethyl)benzene, 2-Bromo-1-chloro-4-(trifluoromethyl)benzene, 3-Bromo-4-chloro-α,α,αa-trifluorotoluene

001639-25G 001639-100G
101782-156EA 21.54 USD
101782-156 101782-158
3-Bromo-4-chlorobenzotrifluoride ≥97%
3-Bromo-4-chlorobenzotrifluoride

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
Boiling Pt: 188 °C (1013 hPa)
Melting Pt: -0.6 °C
Density: 1.743 g/cm³ (25 °C)
MDL Number: MFCD00018093
CAS Number: 454-78-4
EINECS: 207-226-0

Order Now

Specification Test Results

Boiling point 191-192°C
Melting point -23°C
Density 1.726
ND20 1.499

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR