3-Bromo-4-chlorobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

001637-5G 001637-25G 001637-100G
101782-152EA 35.38 USD
101782-152 101782-154 102663-542
3-Bromo-4-chlorobenzoic acid ≥97%
3-Bromo-4-chlorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Melting Pt: 215…216 °C
MDL Number: MFCD00079706
CAS Number: 42860-10-6

Order Now

Specification Test Results

Melting point 215-216°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR