2-Bromo-5-chlorobenzoic acid ≥98%

Supplier: Matrix Scientific

001636-5G 001636-25G 001636-100G
101782-144EA 23.08 USD
101782-144 101782-146 101782-148
2-Bromo-5-chlorobenzoic acid ≥98%
2-Bromo-5-chlorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Melting Pt: 152…154 °C
MDL Number: MFCD00013982
CAS Number: 21739-93-5
EINECS: 244-559-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 152-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR