5-Bromo-2-chlorobenzotrifluoride ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Bromo-4-chloro-3-(trifluoromethyl)benzene, 5-Bromo-2-chlorobenzotrifluoride, 4-Bromo-1-chloro-2-(trifluoromethyl)benzene, 5-Bromo-2-chloro-α,α,α-trifluorotoluene

001634-25G 001634-100G 001634-500G
101782-134EA 20 USD
101782-134 101782-136 101782-138
5-Bromo-2-chlorobenzotrifluoride ≥97%
5-Bromo-2-chlorobenzotrifluoride

Hazard code(s): Irritant

Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
Boiling Pt: 195 °C
Melting Pt: –1.4 °C
Density: 1.7486 g/cm³ (25 °C)
MDL Number: MFCD00000601
CAS Number: 445-01-2
EINECS: 207-149-2

Order Now

Specification Test Results

Density 1.745
ND20 1.507

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR