2-Bromo-4-chloroanisole ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-chloro-1-methoxybenzene

001630-25G 001630-100G
101782-130EA 21.54 USD
101782-130 101782-132
2-Bromo-4-chloroanisole ≥99%
2-Bromo-4-chloroanisole

Hazard code(s): Irritant, Light Sensitive, Store in Freezer

Formula: C₇H₆BrClO
MW: 221.48 g/mol
Boiling Pt: 122 °C (20mm)
Density: 1.61 g/cm³
MDL Number: MFCD00079705
CAS Number: 60633-25-2

Order Now

Specification Test Results

Boiling point 122°C/20mm
Density 1.61

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR