2-Bromo-3-chloro-5-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

001622-1G 001622-5G
101782-108EA 43.08 USD
101782-108 101782-110
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine ≥97%
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆H₂BrClF₃N
Boiling Pt: 80 °C (32mm)
MDL Number: MFCD00153072
CAS Number: 75806-84-7

Order Now

Specification Test Results

Boiling point 80°C/32mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR