1-Bromo-3-chloro-5-fluorobenzene ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Chloro-5-fluorobromobenzene

001620-25G 001620-100G
101782-104EA 18.46 USD
101782-104 101782-106
1-Bromo-3-chloro-5-fluorobenzene ≥98%
1-Bromo-3-chloro-5-fluorobenzene

Hazard code(s): Irritant, Light Sensitive

Formula: C₆H₃BrClF
MW: 209.44 g/mol
MDL Number: MFCD00070747
CAS Number: 33863-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR