2-Bromo-6-chloro-4-(trifluoromethyl)aniline ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-6-Chloro-4-(trifluoromethyl)aniline, 4-Amino-3-bromo-5-chloro-α,α,α-trifluorotoluene, 4-Amino-3-bromo-5-chlorobenzotrifluoride

001615-5G 001615-25G
101782-094EA 21.54 USD
101782-094 101782-096
2-Bromo-6-chloro-4-(trifluoromethyl)aniline ≥98%
2-Bromo-6-chloro-4-(trifluoromethyl)aniline

Hazard code(s): Toxic

Formula: C₇H₄BrClF₃N
MW: 274.47 g/mol
Boiling Pt: 71 - 72 °C (0.25 -rr)
Melting Pt: 29 - 32 °C
MDL Number: MFCD00042180
CAS Number: 109919-26-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 72°C/0.2mm
Melting point 29-32°C
ND20 1.536

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR