2-Amino-5-chloro-2'-fluorobenzophenone
Supplier: Matrix Scientific
Synonyms:
(2-Amino-5-chlorophenyl)(2-fluorophenyl)methanone, 2-Amino-5-chloro-2′-fluorobenzophenone
001606-5G
001606-25G
101782-076EA
32.31
USD
101782-076
101782-078
2-Amino-5-chloro-2'-fluorobenzophenone
2-Amino-5-chloro-2'-fluorobenzophenone
Formula:
C₁₃H₉ClFNO MW: 249.67 g/mol Boiling Pt: 364 °C (1013 hPa) Melting Pt: 94 - 95 °C |
MDL Number:
MFCD00038381 CAS Number: 784-38-3 EINECS: 212-316-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...