1-Bromo-4-butylbenzene ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Bromophenyl)butane

001600-25G 001600-100G
101782-068EA 44.62 USD
101782-068 101782-070
1-Bromo-4-butylbenzene ≥97%
1-Bromo-4-butylbenzene

Hazard code(s): Irritant

Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 125 °C (20mm)
Melting Pt: –26 °C
Density: 1.2347 g/cm³ (20 °C)
MDL Number: MFCD00040934
CAS Number: 41492-05-1

Order Now

Specification Test Results

Boiling point 125°C/20mm
Density 1.208
ND20 1.53

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR