4-Bromo-3-(trifluoromethyl)aniline ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-bromobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-m-toluidine, 5-Amino-2-bromo-α,α,α-trifluorotoluene

001593-25G 001593-100G
101782-054EA 30.77 USD
101782-054 101782-056
4-Bromo-3-(trifluoromethyl)aniline ≥98%
4-Bromo-3-(trifluoromethyl)aniline

MDL: MFCD00007827

Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 81…84 °C (0.5 mmHg)
Melting Pt: 55…56 °C
MDL Number: MFCD00007827
CAS Number: 393-36-2
EINECS: 206-885-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 81-84°C/0.5
Melting point 47-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR