2-Bromo-5-(trifluoromethyl)aniline ≥98%

Supplier: Matrix Scientific
Danger

Synonyms: 3-Amino-4-bromobenzotrifluoride, 6-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-4-bromo-α,α,α-trifluorotoluene, 3-Amino-4-brom-α,α,α-trifluortoluol

001592-5G 001592-25G
101782-050EA 24.62 USD
101782-050 101782-052
2-Bromo-5-(trifluoromethyl)aniline ≥98%
2-Bromo-5-(trifluoromethyl)aniline

Hazard code(s): Irritant

Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 81…82 °C (5mm)
Melting Pt: 47…49 °C
Density: 1.694 g/cm³ (25 °C)
MDL Number: MFCD00007637
CAS Number: 454-79-5
EINECS: 207-227-6
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 81-82°C/5mm
Density 1.675
ND20 1.522

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR