2-Bromo-4-(trifluoromethyl)aniline ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 4-Amino-3-bromobenzotrifluoride, 4-Amino-3-bromo-α,α,α-trifluorotoluene, 2-Bromo-α,α,α-trifluoro-p-toluidine

001591-5G 001591-25G 001591-100G
101782-044EA 24.62 USD
101782-044 101782-046 101782-048
2-Bromo-4-(trifluoromethyl)aniline ≥98% for synthesis
2-Bromo-4-(trifluoromethyl)aniline

Hazard code(s): Irritant

Formula: C₇H₅BrF₃N
Boiling Pt: 110 °C (10mm)
Melting Pt: 26…33 °C
Density: 1.7 g/cm³ (20 °C)
MDL Number: MFCD00042150
CAS Number: 57946-63-1
EINECS: 261-035-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 110°C/10mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR