3-Bromo-5-(trifluoromethyl)aniline ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 3-Amino-5-bromo-α,α,α-trifluorotoluene, 5-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-5-bromobenzotrifluoride

001589-25G 001589-100G
101782-040EA 27.69 USD
101782-040 101782-042
3-Bromo-5-(trifluoromethyl)aniline ≥97% for synthesis
3-Bromo-5-(trifluoromethyl)aniline

Hazard code(s): Irritant

Formula: C₇H₅BrF₃N
Boiling Pt: 220…223 °C
Density: 1.697 g/cm³
MDL Number: MFCD00236205
CAS Number: 54962-75-3
EINECS: 259-412-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR