2-Bromo-6-nitro-4-(trifluoromethyl)aniline ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
4-Amino-3-bromo-5-nitro-α,α,α-trifluorotoluene, 2-Bromo-6-nitro-α,α,α-trifluoro-p-toluidine, 4-Amino-3-bromo-5-nitrobenzotrifluoride
001588-1G
001588-5G
101782-034EA
47.69
USD
101782-034
101782-036
2-Bromo-6-nitro-4-(trifluoromethyl)aniline ≥98%
2-Bromo-6-nitro-4-(trifluoromethyl)aniline
Hazard code(s): Irritant
Formula:
C₇H₄BrF₃N₂O₂ MW: 285.02 g/mol Melting Pt: 68 - 70 °C |
MDL Number:
MFCD00042151 CAS Number: 113170-71-1 |
Specification Test Results
Melting point | 68-70°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...