2-Amino-5-bromo-3-methylpyridine ≥97%
Supplier: Matrix Scientific
|
Warning
|
001585-5G
001585-25G
101782-026EA
21.54
USD
101782-026
101782-028
2-Amino-5-bromo-3-methylpyridine ≥97%
2-Amino-5-bromo-3-methylpyridine
Hazard code(s): Irritant
Formula:
C₆H₇BrN₂ Melting Pt: 92…94 °C |
MDL Number:
MFCD00068232 CAS Number: 3430-21-5 |
Specification Test Results
Melting point | 92-94°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...