2,3,4,5,6-Pentafluorobenzophenone ≥99%

Supplier: Matrix Scientific
Warning

001571-5G 001571-25G
101781-986EA 164.62 USD
101781-986 101781-988
2,3,4,5,6-Pentafluorobenzophenone ≥99%
2,3,4,5,6-Pentafluorobenzophenone

Hazard code(s): Irritant

Formula: C₁₃H₅F₅O
MW: 272.17 g/mol
Boiling Pt: 93 °C (0.2mm)
Melting Pt: 36…39 °C
MDL Number: MFCD00000294
CAS Number: 1536-23-8
EINECS: 216-258-4

Order Now

Specification Test Results

Boiling point 93°C/0.2mm
Melting point 36-39°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR