2,3,4,5,6-Pentafluoroaniline ≥99%

Supplier: Matrix Scientific
Warning

001560-25G 001560-100G
101781-964EA 115.38 USD
101781-964 101781-966
2,3,4,5,6-Pentafluoroaniline ≥99%
2,3,4,5,6-Pentafluoroaniline

Hazard code(s): Irritant

Formula: C₆H₂F₅N
Boiling Pt: 153 °C
Melting Pt: 35…36 °C
Density: 1.744 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007643
CAS Number: 771-60-8
EINECS: 212-234-2

Order Now

Specification Test Results

Boiling point 153°C
Melting point 35-36°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR