trans-3,4-Difluorocinnamic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 3,4-Difluorocinnamic acid

001529-5G 001529-25G
101781-902EA 29.23 USD
101781-902 101781-904
trans-3,4-Difluorocinnamic acid ≥97%
trans-3,4-Difluorocinnamic acid

Hazard code(s): Irritant

Formula: C₉H₆F₂O₂
MW: 184.14 g/mol
Melting Pt: 194…196 °C
MDL Number: MFCD00010320
CAS Number: 112897-97-9

Order Now

Specification Test Results

Melting point 194-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR