trans-2,4-Difluorocinnamic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
(E)-3-(2,4-Difluorophenyl)acrylic acid, 2,4-Difluorocinnamic acid
001526-5G
001526-25G
101781-888EA
38.46
USD
101781-888
101781-890
trans-2,4-Difluorocinnamic acid ≥98%
trans-2,4-Difluorocinnamic acid
Hazard code(s): Irritant
Formula:
C₉H₆F₂O₂ MW: 184.14 g/mol Melting Pt: 215…216 °C |
MDL Number:
MFCD00010317 CAS Number: 94977-52-3 |
Specification Test Results
Melting point | 215-216°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...