trans-2,3-Difluorocinnamic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: (E)-3-(2,3-Difluorophenyl)acrylic acid

001525-1G 001525-5G
101781-884EA 30.77 USD
101781-884 101781-886
trans-2,3-Difluorocinnamic acid ≥97%
trans-2,3-Difluorocinnamic acid

Hazard code(s): Irritant

Formula: C₉H₆F₂O₂
Melting Pt: 198…201 °C
MDL Number: MFCD00061291
CAS Number: 236746-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR