2'-Chloro-6'-fluoroacetophenone

Supplier: Matrix Scientific

Synonyms: 1-(2-Chloro-6-fluorophenyl)ethanone

001511-1G 001511-5G
101781-858EA 30.77 USD
101781-858 101781-860
2'-Chloro-6'-fluoroacetophenone
2'-Chloro-6'-fluoroacetophenone
Formula: C₈H₆ClFO
MW: 172.59 g/mol
MDL Number: MFCD01631390
CAS Number: 87327-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR