2-Methoxy-5-(trifluoromethyl)pyridine ≥98% for synthesis

Supplier: Matrix Scientific

001503-1G 001503-5G 001503-25G
101781-834EA 13.85 USD
101781-834 101781-836 101781-838
2-Methoxy-5-(trifluoromethyl)pyridine ≥98% for synthesis
2-Methoxy-5-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₇H₆F₃NO
MDL Number: MFCD00153203
CAS Number: 175277-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR