2'-Fluoro-5'-(trifluoromethyl)propiophenone ≥97%
Supplier: Matrix Scientific
001498-1G
001498-5G
101781-826EA
30.77
USD
101781-826
101781-828
2'-Fluoro-5'-(trifluoromethyl)propiophenone ≥97%
2'-Fluoro-5'-(trifluoromethyl)propiophenone
Hazard code(s): Irritant
Formula:
C₁₀H₈F₄O MW: 220.17 g/mol |
MDL Number:
MFCD00061260 CAS Number: 207974-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...