(3-Hydroxy-4-methoxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(3-Hydroxy-4-methoxyphenyl)acetic acid

001492-1G 001492-5G
101781-802EA 41.54 USD
101781-802 101781-804
(3-Hydroxy-4-methoxyphenyl)acetic acid ≥98%
(3-Hydroxy-4-methoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
MW: 182.18 g/mol
Melting Pt: 128…130 °C
MDL Number: MFCD00016829
CAS Number: 1131-94-8

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR