3'-Fluoro-4'-methylacetophenone

Supplier: Matrix Scientific

001482-1G 001482-5G
101781-790EA 46.15 USD
101781-790 101781-792
3'-Fluoro-4'-methylacetophenone
3'-Fluoro-4'-methylacetophenone
Formula: C₉H₉FO
MW: 152.17 g/mol
MDL Number: MFCD01631411
CAS Number: 42444-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR