3'-Fluoro-4'-methoxyacetophenone ≥98%

Supplier: Matrix Scientific

001481-25G 001481-100G
101781-786EA 70.77 USD
101781-786 101781-788
3'-Fluoro-4'-methoxyacetophenone ≥98%
3'-Fluoro-4'-methoxyacetophenone

Hazard code(s): Irritant

Formula: C₉H₉FO₂
Boiling Pt: 147 - 148 °C (20 -rr)
Melting Pt: 92 °C
MDL Number: MFCD00026219
CAS Number: 455-91-4
EINECS: 207-253-8

Order Now

Specification Test Results

Boiling point 147-148°C/2
Melting point 90-93°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR