2'-Fluoro-4'-methoxyacetophenone ≥99%
Supplier: Matrix Scientific
001480-5G
001480-25G
101781-782EA
15.38
USD
101781-782
101781-784
2'-Fluoro-4'-methoxyacetophenone ≥99%
2'-Fluoro-4'-methoxyacetophenone
Hazard code(s): Irritant
Formula:
C₉H₉FO₂ MW: 168.17 g/mol Boiling Pt: 243 °C (1013 hPa) Melting Pt: 51…53 °C |
MDL Number:
MFCD00042290 CAS Number: 74457-86-6 |
Specification Test Results
Melting point | 51-53°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...