3'-Chloro-4'-fluoroacetophenone ≥97%

Supplier: Matrix Scientific

001477-5G 001477-25G
101781-768EA 29.23 USD
101781-768 101781-770
3'-Chloro-4'-fluoroacetophenone ≥97%
3'-Chloro-4'-fluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₆ClFO
Boiling Pt: 126…128 °C (15mm)
Melting Pt: 40…42 °C
MDL Number: MFCD00042203
CAS Number: 2923-66-2

Order Now

Specification Test Results

Boiling point 126-128°C/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR