2'-Chloro-4'-fluoroacetanilide ≥99%

Supplier: Matrix Scientific

001475-5G 001475-25G
101781-762EA 24.62 USD
101781-762 101781-764
2'-Chloro-4'-fluoroacetanilide ≥99%
2'-Chloro-4'-fluoroacetanilide

Hazard code(s): Irritant

Formula: C₈H₇ClFNO
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 117…119 °C
MDL Number: MFCD00042594
CAS Number: 399-35-9

Order Now

Specification Test Results

Melting point 117-119°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR