3'-Fluoro-4'-(trifluoromethyl)propiophenone

Supplier: Matrix Scientific

001474-1G 001474-5G
101781-758EA 30.77 USD
101781-758 101781-760
3'-Fluoro-4'-(trifluoromethyl)propiophenone
3'-Fluoro-4'-(trifluoromethyl)propiophenone
Formula: C₁₀H₈F₄O
MW: 220.17 g/mol
MDL Number: MFCD00236306
CAS Number: 237761-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR