(4-Hydroxy-3-nitrophenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

001469-1G 001469-5G
101781-746EA 33.85 USD
101781-746 101781-748
(4-Hydroxy-3-nitrophenyl)acetic acid ≥98%
(4-Hydroxy-3-nitrophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃(OH)(NO₂))CH₂COOH
Melting Pt: 146…148 °C
Storage Temperature: Freezer
MDL Number: MFCD00007122
CAS Number: 10463-20-4
EINECS: 233-948-0

Order Now

Specification Test Results

Melting point 146-148°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR