2-Nitro-4-(trifluoromethyl)anisole ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Methoxy-3-nitrobenzotrifluoride, 1-Methoxy-2-nitro-4-(trifluoromethyl)benzene

001468-25G 001468-100G
101781-742EA 60 USD
101781-742 101781-744
2-Nitro-4-(trifluoromethyl)anisole ≥98%
2-Nitro-4-(trifluoromethyl)anisole

Hazard code(s): Irritant

Formula: C₈H₆F₃NO₃
MW: 221.14 g/mol
Boiling Pt: 90 - 95 °C (0.5 -rr)
Melting Pt: 47 °C
MDL Number: MFCD00007099
CAS Number: 394-25-2
EINECS: 206-891-4
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Boiling point 96-98°C/0.8
Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR