2-Nitro-4-(trifluoromethyl)phenol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-3-nitrobenzotrifluoride

001467-1G 001467-5G
101781-738EA 41.54 USD
101781-738 101781-740
2-Nitro-4-(trifluoromethyl)phenol ≥97%
2-Nitro-4-(trifluoromethyl)phenol

Hazard code(s): Irritant, Stored under Argon

Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
Boiling Pt: 92…94 °C (12mm)
Density: 1.473 g/cm³
MDL Number: MFCD00009791
CAS Number: 400-99-7
EINECS: 206-927-9

Order Now

Specification Test Results

Boiling point 92-94°C/12m
Density 1.473
ND20 1.504

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR