4-Hydroxy-3-nitrobenzyl alcohol ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: (4-Hydroxy-3-nitrophenyl)methanol, 4-(Hydroxymethyl)-2-nitrophenol

001466-5G 001466-25G
101781-734EA 135.38 USD
101781-734 101781-736
4-Hydroxy-3-nitrobenzyl alcohol ≥95% for synthesis
4-Hydroxy-3-nitrobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₇NO₄
Melting Pt: 122 - 123 °C
MDL Number: MFCD00024249
CAS Number: 41833-13-0

Order Now

Specification Test Results

Melting point 97°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR