Homovanillic acid ≥97%

Supplier: Matrix Scientific
Warning

001455-5G 001455-25G
101781-696EA 29.23 USD
101781-696 101781-698
Homovanillic acid ≥97%
Homovanillic acid

Hazard code(s): Irritant

Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 142…145 °C
MDL Number: MFCD00004350
CAS Number: 306-08-1
EINECS: 206-176-7
Merck Index: 13,04761

Order Now

Specification Test Results

Melting point 142-145°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR