2-Methoxy-3-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

001452-5G 001452-25G 001452-100G
101781-686EA 36.92 USD
101781-686 101781-688 101781-690
2-Methoxy-3-(trifluoromethyl)pyridine ≥98%
2-Methoxy-3-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₇H₆F₃NO
MW: 177.13 g/mol
Boiling Pt: 44 °C (15mm)
MDL Number: MFCD00153202
CAS Number: 121643-44-5

Order Now

Specification Test Results

Boiling point 44°C/15mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR