4'-Fluoro-3'-(trifluoromethyl)propiophenone ≥97%

Supplier: Matrix Scientific

001440-1G 001440-5G
101781-658EA 30.77 USD
101781-658 101781-660
4'-Fluoro-3'-(trifluoromethyl)propiophenone ≥97%
4'-Fluoro-3'-(trifluoromethyl)propiophenone

Hazard code(s): Irritant

Formula: C₁₀H₈F₄O
MW: 220.17 g/mol
MDL Number: MFCD00061264
CAS Number: 239107-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR