2'-Fluoro-3'-(trifluoromethyl)propiophenone ≥97%

Supplier: Matrix Scientific

001439-1G 001439-5G
101781-654EA 36.92 USD
101781-654 101781-656
2'-Fluoro-3'-(trifluoromethyl)propiophenone ≥97%
2'-Fluoro-3'-(trifluoromethyl)propiophenone

Hazard code(s): Irritant

Formula: C₁₀H₈F₄O
MW: 220.17 g/mol
Boiling Pt: 128 °C (30mm)
MDL Number: MFCD00061192
CAS Number: 207986-23-0

Order Now

Specification Test Results

Boiling point 128°C/30mm
ND20 1.452

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR