4-Nitro-3-(trifluoromethyl)phenol ≥97%
Supplier: Matrix Scientific
001434-1G
001434-5G
101781-640EA
60
USD
101781-640
101781-642
4-Nitro-3-(trifluoromethyl)phenol ≥97%
4-Nitro-3-(trifluoromethyl)phenol
Hazard code(s): Irritant
Formula:
C₇H₄F₃NO₃ Boiling Pt: 135 - 138 °C (0.01 -rr) Melting Pt: 81 °C |
MDL Number:
MFCD00014714 CAS Number: 88-30-2 EINECS: 201-818-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Boiling point | 135-138°C/0 |
Melting point | 72°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...