2-Thiopheneboronic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
Thiophen-2-ylboronic acid, Thiophene-2-boronic acid, 2-Thienylboronic acid
001426-5G
001426-25G
101781-632EA
21.54
USD
101781-632
101781-634
2-Thiopheneboronic acid ≥97%
2-Thiopheneboronic acid
Hazard code(s): Irritant, Store in Freezer
Formula:
C₄H₅BO₂S MW: 127.96 g/mol Melting Pt: 138…140 °C |
MDL Number:
MFCD00151850 CAS Number: 6165-68-0 |
Specification Test Results
Melting point | 138-140°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...