3-Hydroxy-2,4,6-tribromobenzoic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2,4,6-Tribromo-3-hydroxybenzoic acid

001412-5G 001412-25G
101781-602EA 30.77 USD
101781-602 101781-604
3-Hydroxy-2,4,6-tribromobenzoic acid ≥98%
3-Hydroxy-2,4,6-tribromobenzoic acid

MDL: MFCD00055557

Formula: C₇H₃Br₃O₃
MW: 374.81 g/mol
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 146 °C
MDL Number: MFCD00055557
CAS Number: 14348-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR