5'-Fluoro-2'-hydroxyacetophenone ≥98%
Supplier: Matrix Scientific
001407-25G
001407-100G
101781-586EA
44.62
USD
101781-586
101781-588
5'-Fluoro-2'-hydroxyacetophenone ≥98%
5'-Fluoro-2'-hydroxyacetophenone
Hazard code(s): Irritant
Formula:
C₈H₇FO₂ Boiling Pt: 182 °C (1013 hPa) Melting Pt: 56…58 °C |
MDL Number:
MFCD00011668 CAS Number: 394-32-1 EINECS: 206-893-5 |
Specification Test Results
Melting point | 56-58°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...