4'-Chloro-2'-fluoroacetanilide ≥98%

Supplier: Matrix Scientific

Synonyms: N-(4-Chloro-2-fluorophenyl)acetamide

001405-25G 001405-100G
101781-576EA 30.77 USD
101781-576 101781-578
4'-Chloro-2'-fluoroacetanilide ≥98%
4'-Chloro-2'-fluoroacetanilide

Hazard code(s): Irritant

Formula: C₈H₇ClFNO
MW: 187.6 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 156 - 158 °C
MDL Number: MFCD00042520
CAS Number: 59280-70-5

Order Now

Specification Test Results

Melting point 165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR