4-Hydroxy-1-naphthaldehyde for synthesis

Supplier: Matrix Scientific

001398-1G 001398-5G
101781-556EA 67.69 USD
101781-556 101781-558
4-Hydroxy-1-naphthaldehyde for synthesis
4-Hydroxy-1-naphthaldehyde

Hazard code(s): Irritant

Formula: C₁₁H₈O₂
Melting Pt: 176…178 °C
MDL Number: MFCD00053486
CAS Number: 7770-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR