Decafluoro-4,6-bis(trifluoromethyl)cyclohexane ≥80%, tech.

Supplier: Matrix Scientific
Warning

Synonyms: Perfluoro-1,3-dimethylcyclohexane, Perfluoro(1,3-dimethylcyclohexane)

001392-25G 001392-100G
101781-550EA 92.31 USD
101781-550 101781-552
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane ≥80%, tech.
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane

RTECS: GV4990000 TSCA: T MDL: MFCD00001469


  • RTECS: GV4990000
  • TSCA: T

Formula: C₈F₁₆
MW: 400.06 g/mol
MDL Number: MFCD00001469
CAS Number: 335-27-3
EINECS: 206-386-9

Order Now

Specification Test Results

Boiling point 101-102°C
Melting point -55°C
Density 1.828
ND20 1.3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR